Merck
Physicochemical study of the acetonitrile insertion into polypyrrole films.
The Journal of chemical physics (2010-04-22)
S D Oliveira Costa, A J Fernández Romero, J J López Cascales
ABSTRAKT
A study by molecular dynamics (MD) simulation of the acetonitrile diffusion into a polypyrrole film was carried out with atomic detail in a 0.1N lithium perchlorate solution. From the simulated trajectories, the acetonitrile behavior was estimated from bulk solution to the interior of the polypyrrole film, across the polypyrrole/solution interface, for a neutral (reduced) and charged (oxidized) state of the polymer. Among other properties, the translational diffusion coefficient and rotational relaxation time of the acetonitrile were calculated, where a diminution in the translational diffusion coefficient was measured in the interior of the polypyrrole matrix compared to bulk, independently of the oxidation state of the polymer, in contrast with the behavior of the rotational relaxation time that increases from bulk to the interior of the polymer for both oxidation states. In addition, the difference of free energy DeltaG associated to the acetonitrile penetration into the polymer was calculated. From the results, it was evidenced that the scarce affinity of acetonitrile to diffuse into the polymer in its reduced state is related with the positive uniform difference of free energy DeltaG approximately 20 kJ/mol, while in the oxidized state, an important free energy barrier of DeltaG approximately 10 kJ/mol has to pass trough for reaching stable sites inside the polymer with values of DeltaG up to -10 kJ/mol.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Lithium perchlorate, 99.99% trace metals basis
Sigma-Aldrich
Lithium perchlorate, purum p.a., ≥98.0% (calc. based on dry substance, T), powder
Sigma-Aldrich
Lithium perchlorate, ACS reagent, ≥95.0%
Sigma-Aldrich
Lithium perchlorate, battery grade, dry, 99.99% trace metals basis