Merck
Monitoring of progestins: development of immunochemical methods for purification and detection of levonorgestrel.
Analytica chimica acta (2010-04-27)
Moran Shalev, Michal Bardugo, Ayelet Nudelman, Alex Krol, Haim Schlesinger, Alisa Bronshtein, Miriam Altstein
ABSTRAKT
A polyclonal antibody (Ab) for the progestin levonorgestrel (LNG) was generated, and immunochemical assays for its detection, clean-up and concentration were developed. A highly specific microplate diagnostic assay for the detection of LNG was developed that used the enzyme linked immunosorbent assay (ELISA) method. The LNG ELISA developed was sensitive and reproducible; it exhibited I(50) and I(20) values of 3.3+/-1.8 ng mL(-1) and 0.6+/-0.4 ng mL(-1), respectively, and the Abs did not cross react with any of the tested steroid hormones. The above Abs were used to develop a sol-gel-based immunoaffinity purification (IAP) method for concentration and clean-up of LNG that is compatible with subsequent immunochemical or instrumental chemical analytical procedures, such as liquid chromatography followed by mass spectrometry (LC-MS/MS). Development of the columns included successful entrapment of Abs within a tetramethoxysilane (TMOS)-based SiO(2) polymer network. The Abs could bind the free analyte from solution, and the bound analyte could be easily eluted from the sol-gel matrix at high recoveries. The Ab selectivity towards the antigen was high, in both ELISA and the sol-gel columns, but the entrapped Abs cross-reacted with two other steroid hormones--ethynylestradiol (EE2) and nortestosterone (NT) - which share similar epitopes with LNG, despite the lack of cross reactivity in the ELISA. The validity of the method was investigated by LC-MS/MS and a good analytical correlation was obtained.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Tetramethyl orthosilicate, 98%
Sigma-Aldrich
Tetramethyl orthosilicate, ≥99%
Sigma-Aldrich
Tetramethyl orthosilicate, purum, ≥98.0% (GC)