Merck
Indirect determination of isocyanates by gas chromatography.
Journal of chromatography (1991-01-11)
C B Fanska, T J Byerley, J D Eick
ABSTRAKT
A gas chromatographic method was developed to determine the purity of synthesized isocyanate monomers, specifically isocyanatoacrylates, and to determine the isocyanate content of synthesized polymers and prepolymers. The method is a modification of an ASTM procedure in which an isocyanate is allowed to react with excess di-n-butylamine. In the ASTM method, the amount of isocyanate present is calculated indirectly from the amount of unreacted di-n-butylamine, determined by back titration with standard hydrochloric acid. Determination of the excess di-n-butylamine by the gas chromatographic method developed has the advantages of providing better precision and requiring less sample than the titrimetric method. The two methods were compared using phenyl isocyanate as a model test compound. A synthesized monomer, methyl alpha-isocyanatoacrylate, was also analyzed, for comparison by both methods.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Phenyl isocyanate, derivatization grade (HPLC), LiChropur, ≥99.0% (GC)
Sigma-Aldrich
Phenyl isocyanate, ≥98%