Merck
Statins: in the beginning.
The journal of the Royal College of Physicians of Edinburgh (2010-06-01)
K S Lyons, M Harbinson
ABSTRAKT
Since the first human trial of a hydroxymethylglutaryl-coenzyme A (HMG CoA) reductase inhibitor in 1978, the growth in importance of this drug class, both financially and medically, has been staggering. The aim of this paper is to summarise how this drug class was developed, highlighting the role ofAkira Endo.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Mevastatin, ≥98% (HPLC), powder or crystals