Merck
Low temperature synthesis of ZnS and CdZnS shells on CdSe quantum dots.
Nanotechnology (2010-06-03)
Huiguang Zhu, Arjun Prakash, Denise N Benoit, Christopher J Jones, Vicki L Colvin
ABSTRAKT
Methods for synthesizing quantum dots generally rely on very high temperatures to both nucleate and grow core and core-shell semiconductor nanocrystals. In this work, we generate highly monodisperse ZnS and CdZnS shells on CdSe semiconductor nanocrystals at temperatures as low as 65 degrees C by enhancing the precursor solubility. Relatively small amounts of trioctylphosphine and trioctylphosphine oxide have marked effects on the solubility of the metal salts used to form shells; their inclusion in the precursor solutions, which use thiourea as a sulfur source, can lead to homogeneous and fully dissolved solutions. Upon addition to suspensions of quantum dot cores, these precursors deposit as uniform shells; the lowest temperature for shell growth (65 degrees C) yields the thinnest shells (d < 1 nm) while the same process at higher temperatures (180 degrees C) forms thicker shells (d approximately 1-2 nm). The growth of the shell structures, average particle size, size distribution, and shape were examined using optical spectroscopy, transmission electron microscopy, x-ray diffraction, and transmittance small angle x-ray scattering. The photoluminescence quantum yield (QY) of the as-prepared CdSe/ZnS quantum dots ranged from 26% to 46% as compared to 10% for the CdSe cores. This method was further generalized to CdZnS shells by mixing cadmium and zinc acetate precursors. The CdSe/CdZnS nanocrystals have a thicker shell and higher QY (40% versus 36%) as compared to the CdSe/ZnS prepared under similar conditions. These low temperature methods for shell growth are readily amenable to scale-up and can provide a route for economical and less energy intensive production of quantum dots.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
1-Octadecene, ≥95.0% (GC)
Sigma-Aldrich
1-Octadecene, technical grade, 90%
Supelco
1-Octadecene, analytical standard, ≥99.0% (GC)