Merck
Preparation and characterization of ketoprofen-loaded solid lipid nanoparticles made from beeswax and carnauba wax.
Nanomedicine : nanotechnology, biology, and medicine (2010-07-06)
Soheila Kheradmandnia, Ebrahim Vasheghani-Farahani, Mohsen Nosrati, Fatemeh Atyabi
ABSTRAKT
Solid lipid nanoparticles (SLNs) have been proposed as suitable colloidal carriers for delivery of drugs with limited solubility. Ketoprofen as a model drug was incorporated into SLNs prepared from a mixture of beeswax and carnauba wax using Tween 80 and egg lecithin as emulsifiers. The characteristics of the SLNs with various lipid and surfactant composition were investigated. The mean particle size of drug-loaded SLNs decreased upon mixing with Tween 80 and egg lecithin as well as upon increasing total surfactant concentration. SLNs of 75 ± 4 nm with a polydispersity index of 0.2 ± 0.02 were obtained using 1% (vol/vol) mixed surfactant at a ratio of 60:40 Tween 80 to egg lecithin. The zeta potential of these SLNs varied in the range of -15 to -17 (mV), suggesting the presence of similar interface properties. High drug entrapment efficiency of 97% revealed the ability of SLNs to incorporate a poorly water-soluble drug such as ketoprofen. Differential scanning calorimetry thermograms and high-performance liquid chromatographic analysis indicated the stability of nanoparticles with negligible drug leakage after 45 days of storage. It was also found that nanoparticles with more beeswax content in their core exhibited faster drug release as compared with those containing more carnauba wax in their structure.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Carnauba wax No. 1 yellow, refined