Merck
Studies on the mode of action of tolnaftate in Microsporum gypseum.
Journal of medical and veterinary mycology : bi-monthly publication of the International Society for Human and Animal Mycology (1991-01-01)
M P Gupta, N Kapur, I Bala, G K Khuller
ABSTRAKT
Studies were performed on the mode of action of tolnaftate and resistance to this drug in Microsporum gypseum. Cells grown in the presence of tolnaftate (at the IC 50) showed a reduced content of total phospholipids and sterols whereas there was an increase in total RNA content. Incubation of cells with tolnaftate (at 10 x MIC), followed by addition of different macromolecule precursors revealed inhibition of the biosynthesis of all macromolecules except for RNA. The activity of membrane-bound enzymes did not change on treatment with tolnaftate (10 x MIC) whereas an increase in the leakage of intracellular 32P was observed. The content of total phospholipids was higher in tolnaftate-resistant cells, whereas the content of total sterols, DNA, RNA and protein was comparable to that of susceptible cultures. Activity of phosphodiesterase decreased and 5'-nucleotidase increased in tolnaftate-resistant cells. Our results suggest that the antifungal activity of tolnaftate is due to differential action on various targets site(s) which are modified in strains resistant to the drug.