Merck
  • Using a first-derivative spectrophotometric method for simultaneous determination of Fe(II) and Ni(II) in mineral vitamins by interaction with 2,4-diamino-5-(2-hydroxy-5-nitrophenylazo)benzenesulfonic acid sodium salt in the presence of Tween 20.
Using a first-derivative spectrophotometric method for simultaneous determination of Fe(II) and Ni(II) in mineral vitamins by interaction with 2,4-diamino-5-(2-hydroxy-5-nitrophenylazo)benzenesulfonic acid sodium salt in the presence of Tween 20.
Journal of AOAC International (2010-07-16)
Elham Y Hashem, Mohamed M Seleim, Ahmed M El-Zohry
ABSTRAKT
A highly selective and sensitive derivative spectrophotometric method has been developed for the determination of Fe(II) and Ni(II) in different mineral vitamins. The method is based on the formation of binary complexes of Fe(II) and Ni(II) with 2,4-diamino-5-(2-hydroxy-5-nitrophenylazo) benzenesulfonic acid sodium salt (Mordant Brown 33) at pH 5.6 with Tween 20. At lambda(max) 512 and 493 nm, the molar absorbtivity was 2.09 x 10(4) and 0.58 x 10(4) L/mol cm for Fe(II) and Ni(II), respectively. Fe(II) can be determined in the range 0.55-2.79 microg/mL in the presence of 2.92 microg/mL Ni, and Ni(II) can be determined in the range 1.17-3.5 microg/mL in the presence of 2.79 microg/mL Fe(II) in the presence of Tween 20 (4%). The detection limits were 9.0 and 15.0 ng/mL for Fe(II) and Ni(II), respectively. The effect of foreign ions was elucidated. The RSD values were, in all instances, less than 1.3%. The proposed method was successfully applied for the simultaneous determination of Fe(II) and Ni(II) in different mineral vitamins.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sodium benzenesulfonate, 97%
Sigma-Aldrich
Benzenesulfonic acid, 98.0% (T)
Sigma-Aldrich
Benzenesulfonic acid, technical grade, 90%