Merck
  • Catalytic and regioselective ring expansion of arylcyclobutanones with trimethylsilyldiazomethane. Ligand-dependent entry to beta-ketosilane or enolsilane adducts.
Catalytic and regioselective ring expansion of arylcyclobutanones with trimethylsilyldiazomethane. Ligand-dependent entry to beta-ketosilane or enolsilane adducts.
Organic letters (2010-07-31)
Jennifer A Dabrowski, David C Moebius, Andrew J Wommack, Anne F Kornahrens, Jason S Kingsbury
ABSTRAKT
Divergent reactivity is uncovered in the homologation of arylcyclobutanones with trimethylsilyldiazomethane. With Sc(OTf)(3) as catalyst, enolsilanes are obtained with a high preference for methylene migration. By contrast, Sc(hfac)(3) gives beta-ketosilanes with both regio- and diastereocontrol. Each adduct affords the cyclopentanone upon hydrolysis.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Cyclopentanone, ReagentPlus®, ≥99%
Supelco
Cyclopentanone, analytical standard
Sigma-Aldrich
Cyclopentanone, ≥99%, FG