Merck
4-Methoxyanilinium tetrafluoroborate-dibenzo-18-crown-6 (1/1).
Acta crystallographica. Section C, Crystal structure communications (2010-08-04)
Xue-qun Fu, Yong-le Yang, Qiong Ye
ABSTRAKT
In the structure of the complex of dibenzo-18-crown-6 [systematic name: 2,5,8,15,18,21-hexaoxatricyclo[20.4.0.0(9,14)]hexacosa-1(26),9,11,13,22,24-hexaene] with 4-methoxyanilinium tetrafluoroborate, C(7)H(10)NO(+) x BF(4)(-) x C(20)H(24)O(6), the protonated 4-methoxyanilinium (MB-NH(3)(+)) cation forms a 1:1 supramolecular rotator-stator complex with the dibenzo-18-crown-6 molecule via N-H...O hydrogen bonds. The MB-NH(3)(+) group is attached from the convex side of the bowl-shaped crown, in contrast with similar ammonium cations that nest in the curvature of the bowl. The cations are associated via C-H...pi interactions, while the cations and anions are linked by weak C-H...F hydrogen bonds, forming cation-crown-anion chains parallel to [011].
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
p-Anisidine, 99%
Sigma-Aldrich
Dibenzo-18-crown-6, 98%