Merck
  • A multicenter, randomized controlled clinical study on biapenem and imipenem/cilastatin injection in the treatment of respiratory and urinary tract infections.
A multicenter, randomized controlled clinical study on biapenem and imipenem/cilastatin injection in the treatment of respiratory and urinary tract infections.
Chemotherapy (2010-08-18)
Bei Jia, Ping Lu, Wenxiang Huang, Chongzhi Li, Ailong Huang, Xiangdong Zhou, Weili Zhang, Guoming Wu, Genfu Zhang
ABSTRAKT
Biapenem is an injectable carbapenem antibiotic. A clinical study was designed to evaluate its efficacy and safety in the treatment of respiratory and urinary infections compared to imipenem/cilastatin. A total of 216 patients with respiratory or urinary tract infections were enrolled into this multicenter, open-label, randomized controlled clinical study. Each patient was randomly assigned to either the treatment or control group; 106 patients in each group were included in the ITT analyses. The patients were given biapenem 300 mg or imipenem/cilastatin 500 mg/500 mg two or three times daily, i.v. g.t.t. for 7-14 days according to their conditions. The cure and effective rates were 67.92 and 88.68% in the biapenem group and 76.02 and 93.40% in the imipenem/cilastatin group, the bacterial eradication rates were 93.83 and 98.82%, and the adverse-event rates were 6.54 and 7.41%, respectively. There were no significant differences between the two groups (p > 0.05). Biapenem is as effective and well-tolerated as imipenem/cilastatin for the treatment of intermediate and severe bacterial infections.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Biapenem, ≥98% (HPLC)