Merck
Synthesis of a mesoporous single crystal Ga2O3 nanoplate with improved photoluminescence and high sensitivity in detecting CO.
Chemical communications (Cambridge, England) (2010-08-18)
Shicheng Yan, Lijuan Wan, Zhaosheng Li, Yong Zhou, Zhigang Zou
ABSTRAKT
A new approach is proposed to synthesize a mesoporous single crystal Ga(2)O(3) nanoplate by heating a single crystal nanoplate of GaOOH, which involves an ion exchange between KGaO(2) and CH(3)COOH at room temperature for the formation of GaOOH and pseudomorphic and topotactic phase transformation from GaOOH to Ga(2)O(3).
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Gallium(III) oxide, ≥99.99% trace metals basis