Merck
LC for analysis of two sustained-release mixtures containing cough cold suppressant drugs.
Journal of chromatographic science (2010-09-09)
Alaa El-Gindy, Shehab Sallam, Randa A Abdel-Salam
ABSTRAKT
A liquid chromatographic method was applied for the analysis of two sustained-release mixtures containing dextromethorphane hydrobromide, carbinoxamine maleate with either phenylephrine hydrochloride in pharmaceutical capsules (Mix 1) or phenyl-propanolamine, methylparaben, and propylparaben, which bonds as a drug base to ion exchange resin in pharmaceutical syrup (Mix 2). The method was used for their simultaneous determination using a CN column with a mobile phase consisting of acetonitrile-12 mM ammonium acetate in the ratio of 60:40 (v/v, pH 6.0) for Mix 1 and 45:55 (v/v, pH 6.0) for Mix 2.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Propylparaben, Pharmaceutical Secondary Standard; Certified Reference Material
Propyl parahydroxybenzoate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Propyl 4-hydroxybenzoate, ≥99%
Sigma-Aldrich
Propyl 4-hydroxybenzoate, BioXtra
Sigma-Aldrich
Propyl 4-hydroxybenzoate, tested according to Ph. Eur.