Merck
Preparation of poly(2-amino thiophenol) nanodiscs by a "combined hard-soft template" approach and characterization.
Journal of colloid and interface science (2010-09-21)
Shanmugasundaram Komathi, Subramanian Palaniappan, Paramasivam Manisankar, Anantha Iyengar Gopalan, Kwang-Pill Lee
ABSTRAKT
A seed induced chemical oxidative polymerization was used for the preparation of pure poly(2-amino thiophenol) nanodiscs (P2AT-NDs (P)). Two templates, (hard (MCM-41) and soft (β-napthalene sulfonic acid), were utilized for the preparation of the seed, P2AT nanostructures loaded MCM-41. The field emission scanning electron microscopy reveals nanodisc morphology for P2AT (P). X-ray diffraction, current-potential characteristics and electrochemical impedance spectroscopy were used to evaluate the physicochemical properties of P2AT-ND (P). The P2AT-ND (P) exhibits semicrystalline behaviour, good electron transport and lesser charge transfer resistance at the interface as compared to simple P2AT prepared by conventional chemical route.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
2-Aminothiophenol, technical grade, 90%
Sigma-Aldrich
2-Aminothiophenol, 99%