Merck
Development of drug-in-adhesive transdermal patch for α-asarone and in vivo pharmacokinetics and efficacy evaluation.
Drug delivery (2010-10-14)
Ying Hu, Yao-Yao Wu, Xiao-Jing Xia, Zheng Wu, Wen-Quan Liang, Jian-Qing Gao
ABSTRAKT
A transdermal drug delivery system has been reported that can increase the bioavailability, reduce the administration duration, and maintain the concentration of drug in blood. In the present study, drug-in-adhesive transdermal patches of α-asarone using Eudragit E100 as pressure-sensitive adhesives and oleic acid plus isopropyl myristate as penetration co-enhancers were developed. In vitro permeation, in vivo pharmacokinetics in rabbits, and efficacy in asthmatic rats were evaluated. The results showed that co-enhancers could induce a synergistic effect on α-asarone permeability. In vivo study suggested that the patch can keep a relatively certain blood level of drug within 10-30 h in rabbits. Furthermore, the patch with the size of 4 cm² containing drug 3 mg/cm² showed a noticeable treating effect on asthmatic rats which is equivalent to the effect of dexamethasone, while avoiding the side-effect induced by the corticorsteroid. This suggests that the drug-in-adhesive transdermal patch is a promising delivery system containing α-asarone to be used for asthma treatment.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
cis-2,4,5-Trimethoxy-1-propenylbenzene, 70%