Merck
  • Chemical genetics approach to identify new small molecule modulators of cell growth by phenotypic screening of Saccharomyces cerevisiae strains with a library of morpholine-derived compounds.
Chemical genetics approach to identify new small molecule modulators of cell growth by phenotypic screening of Saccharomyces cerevisiae strains with a library of morpholine-derived compounds.
Organic & biomolecular chemistry (2010-10-16)
Andrea Trabocchi, Irene Stefanini, Manfredi Morvillo, Leonardo Ciofi, Duccio Cavalieri, Antonio Guarna
ABSTRAKT
A chemical genetics approach has been applied in the screening of yeast deletants strains with a pool of morpholine-derived compounds in order to identify candidate small molecules able to produce phenotypic effects on yeast cells. The analysis of the effects of structurally diverse molecules towards cell growth rate in both exponential and stationary phases provides a tool to select candidate compounds for subsequent assays to identify new chemical entities as chemical probes for drug discovery.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Morpholine, ACS reagent, ≥99.0%
Sigma-Aldrich
Morpholine, ReagentPlus®, ≥99%
Supelco
Morpholine, analytical standard
Sigma-Aldrich
Morpholine, purified by redistillation, ≥99.5%