Merck
  • Simultaneous determination of NOGE-related and BADGE-related compounds in canned food by ultra-performance liquid chromatography-tandem mass spectrometry.
Simultaneous determination of NOGE-related and BADGE-related compounds in canned food by ultra-performance liquid chromatography-tandem mass spectrometry.
Analytical and bioanalytical chemistry (2010-10-19)
Hong Zhang, Ming Xue, Yanyan Zou, Zhiyuan Dai, Kunhui Lin
ABSTRAKT
An improved analytical method enabling rapid and accurate determination and identification of bisphenol F diglycidyl ether (novolac glycidyl ether 2-ring), novolac glycidyl ether 3-ring, novolac glycidyl ether 4-ring, novolac glycidyl ether 5-ring, novolac glycidyl ether 6-ring, bisphenol A diglycidyl ether, bisphenol A (2,3-dihydroxypropyl) glycidyl ether, bisphenol A (3-chloro-2-hydroxypropyl) glycidyl ether, bisphenol A bis(3-chloro-2-hydroxypropyl) ether, and bisphenol A (3-chloro-2-hydroxypropyl) (2,3-dihydroxypropyl) ether in canned food and their contact packaging materials has been developed by using, for the first time, ultra-performance liquid chromatography coupled with tandem mass spectrometry. After comparison of electrospray ionization and atmospheric pressure chemical ionization in positive and negative-ion modes, tandem mass spectrometry with positive electrospray ionization was chosen to carry out selective multiple reaction monitoring analysis of novolac glycidyl ethers, bisphenol A diglycidyl ether, and its derivatives. The analysis time is only 5.5 min per run. Limits of detection varied from 0.01 to 0.20 ng g(-1) for the different target compounds on the basis of a signal-to-noise ratio (S/N) = 3; limits of quantitation were from 0.03 to 0.66 ng g(-1). The relative standard deviation for repeatability was <8.01%. Analytical recovery ranged from 87.60 to 108.93%. This method was successfully applied to twenty samples of canned food and their contact packaging materials for determination of migration of NOGE, BADGE, and their derivatives from can coatings into food.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
D.E.R. 332, used as embedding medium
Sigma-Aldrich
Bisphenol A diglycidyl ether
Supelco
Bisphenol A diglycidyl ether, analytical standard