Merck
Copper-catalyzed decarboxylative cross-coupling of alkynyl carboxylic acids with aryl halides.
Chemical communications (Cambridge, England) (2010-11-05)
Dongbing Zhao, Chao Gao, Xiaoyu Su, Yunqing He, Jingsong You, Ying Xue
ABSTRAKT
The copper-catalyzed decarboxylative reactions of alkynyl carboxylic acids with aryl halides were performed under relatively mild reaction conditions. Benzofurans could be further prepared smoothly by a one-pot domino protocol on the basis of decarboxylative cross-coupling of 2-iodophenol.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
2-Iodophenol, 98%
Sigma-Aldrich
Phenylpropiolic acid, 99%