Merck
Sustained release of nucleic acids from polymeric nanoparticles using microemulsion precipitation in supercritical carbon dioxide.
Chemical communications (Cambridge, England) (2010-11-06)
Jun Ge, Gunilla B Jacobson, Tatsiana Lobovkina, Krister Holmberg, Richard N Zare
ABSTRAKT
A general approach for producing biodegradable nanoparticles for sustained nucleic acid release is presented. The nanoparticles are produced by precipitating a water-in-oil microemulsion in supercritical CO(2). The microemulsion consists of a transfer RNA aqueous solution (water phase), dichloromethane containing poly(l-lactic acid)-poly(ethylene glycol) (oil phase), the surfactant n-octyl β-D-glucopyranoside, and the cosurfactant n-butanol.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Octyl β-D-1-thioglucopyranoside, ≥98.0% (GC)