Merck
Helical polymers based on intramolecularly hydrogen-bonded aromatic polyamides.
Chemical communications (Cambridge, England) (2010-11-09)
Yi-Xuan Lu, Zhu-Ming Shi, Zhan-Ting Li, Zhibin Guan
ABSTRAKT
Inspired by arylamide-based oligomeric foldermers that are stabilized by intramolecular hydrogen bonding, a series of polyamides with intramolecular hydrogen-bonding motifs were synthesized via polycondensation reactions. These polymers can fold into helical conformation different from their linear control. The chirality of helical conformation can further be tuned via acid-base complexation using chiral residues.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Triphenyl phosphite, 97%