Merck
Advantages of isothermal titration calorimetry for xylanase kinetics in comparison to chemical-reducing-end assays.
Analytical biochemistry (2010-11-16)
Martin J Baumann, Leigh Murphy, Nina Lei, Kristian B R M Krogh, Kim Borch, Peter Westh
ABSTRAKT
In lignocellulosic raw materials for biomass conversion, hemicelluloses constitute a substantial fraction, with xylan being the primary part. Although many pretreatments reduce the amount or change the distribution of xylan, it is important to degrade residual xylan so as to improve the overall yield. Typically, xylanase reaction rates are measured in stopped assays by chemical quantification of the reducing ends. With isothermal titration calorimetry (ITC), the heat flow of the hydrolysis can be measured in continuous fashion, with the reaction rate being directly proportional to the heat flow. Reaction enthalpies for carbohydrate hydrolysis are typically below 5kJ/mol, which is the limiting factor for straight forward calorimetric quantification of enzymatic reaction rates using current ITC technology. To increase the apparent reaction enthalpy, we employed a subsequent oxidation of hydrolysis products by carbohydrate oxidase and catalase. Here we show that the coupled assay with carbohydrate oxidase and catalase can be used to measure enzyme kinetics of a GH10 xylanase from Aspergillus aculeatus on birch xylan and wheat arabinoxylan. Results are discussed in the light of a critical analysis of the sensitivity of four chemical-reducing-end quantification methods using well-characterized substrates.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
4-Hydroxybenzhydrazide, ≥97%