Merck
Rapid, quantitative, and site specific synthesis of biomolecular radicals from a simple photocaged precursor.
Chemical communications (Cambridge, England) (2011-01-25)
Tony Ly, Xing Zhang, Qingyu Sun, Benjamin Moore, Yuanqi Tao, Ryan R Julian
ABSTRAKT
Novel p-iodobenzoate-based labelling reagents are shown to be effective photocaged precursors for synthesizing biomolecular radicals site-selectively in the gaseous and condensed phases. In vacuo, a single pulse of UV photons (266 nm) is sufficient to quantitatively photolyse the C-I bond. In aqueous solutions, the photolysis half-life is estimated to be 2.5 minutes when irradiating with a 15 W compact fluorescent lamp (254 nm).
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Iodobenzene, 98%