Merck
I2 molecular elimination in single-photon dissociation of CH2I2 at 248 nm by using cavity ring-down absorption spectroscopy.
The Journal of chemical physics (2011-01-26)
Shun-Yi Chen, Po-Yu Tsai, Hsiang-Chin Lin, Chia-Ching Wu, King-Chuen Lin, B J Sun, A H H Chang
ABSTRAKT
Following single-photon dissociation of CH(2)I(2) at 248 nm, I(2) molecular elimination is detected by using cavity ring-down absorption spectroscopy. The technique comprises two laser beams propagating in a perpendicular configuration, in which a tunable laser beam along the axis of the ring-down cell probes the I(2) fragment in the B (3)Π(ou)(+) - X (1)Σ(g)(+) transition. The nascent vibrational populations for v = 0, 1, and 2 levels are obtained with a population ratio of 1:(0.65 ± 0.10):(0.30 ± 0.05), corresponding to a Boltzmann-like vibrational temperature of 544 ± 73 K. The quantum yield of the ground state I(2) elimination reaction is determined to be 0.0040 ± 0.0025. With the aid of ab initio potential energy calculations, the pathway of molecular elimination is proposed on the energetic ground state CH(2)I(2) via internal conversion, followed by asynchronous three-center dissociation. A positive temperature effect supports the proposed mechanism.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Diiodomethane, ReagentPlus®, 99%, contains copper as stabilizer