Merck
o-Semiquinonato and o-iminosemiquinonato rhodium complexes. EPR study of the reactions in coordination sphere of rhodium.
Journal of magnetic resonance (San Diego, Calif. : 1997) (2011-02-11)
M P Bubnov, I A Teplova, K A Kozhanov, G A Abakumov, V K Cherkasov
ABSTRAKT
The number of dicarbonyl-o-semiquinonato (o-iminosemiquinonato) rhodium complexes was isolated and characterized. Some reactions of these compounds with tertiary phosphines (substitution, addition) were studied by EPR in solution using o-semiquinones (o-iminosemiquinones) as spin labels.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Triphenylphosphine, ReagentPlus®, 99%
Sigma-Aldrich
Triphenylphosphine, ≥95.0% (GC)