Merck
Disproportionation for growing copper nanowires and their controlled self-assembly facilitated by ligand exchange.
Chemistry (Weinheim an der Bergstrasse, Germany) (2011-02-11)
Enyi Ye, Shuang-Yuan Zhang, Shuhua Liu, Ming-Yong Han
ABSTRAKT
The coating makes the wire bundle: High-quality free-standing copper nanowires have been successfully produced by disproportionation of Cu(+) in oleylamine. This provides an effective way to prepare high-quality copper nanowires, but also enriches synthetic routes to other nanostructures. These copper nanowires can self-assemble by surface ligand exchange of oleylamine with trioctylphosphine.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Trioctylphosphine oxide, technical grade, 90%
Sigma-Aldrich
Trioctylphosphine oxide, ReagentPlus®, 99%
Sigma-Aldrich
Oleylamine, technical grade, 70%
Sigma-Aldrich
Trioctylphosphine oxide, for extraction analysis, ≥98.5%