Merck
Functional characterization of a special thermophilic multifunctional amylase OPMA-N and its N-terminal domain.
Acta biochimica et biophysica Sinica (2011-03-01)
Fan Li, Xuejun Zhu, Yanfei Li, Hao Cao, Yingjiu Zhang
ABSTRAKT
A gene encoding a special thermophilic multifunctional amylase OPMA-N was cloned from Bacillus sp. ZW2531-1. OPMA-N has an additional 124-residue N-terminal domain compared with typical amylases and forms a relatively independent domain with a β-pleated sheet and random coil structure. Here we reported an unusual substrate and product specificities of OPMA-N and the impact of the additional N-terminal domain (1-124 aa) on the function and properties of OPMA-N. Both OPMA-N (12.82 U/mg) and its N-terminal domain-truncated ΔOPMA-N (12.55 U/mg) only degraded starch to produce oligosaccharides including maltose, maltotriose, isomaltotriose, and isomaltotetraose, but not to produce glucose. Therefore, the N-terminal domain did not determine its substrate and product specificities that were probably regulated by its C-terminal β-pleated sheet structure. However, the N-terminal domain of OPMA-N seemed to modulate its catalytic feature, leading to the production of more isomaltotriose and less maltose, and it seemed to contribute to OPMA-N's thermostability since OPMA-N showed higher activity than ΔOPMA-N in a temperature range from 40 to 80°C and the half-life (t(1/2)) was 5 h for OPMA-N and 2 h for ΔOPMA-N at 60°C. Both OPMA-N and ΔOPMA-N were Ca(2+)-independent, but their activities could be influenced by Cu(2+), Ni(2+), Zn(2+), EDTA, SDS (1 mM), or Triton-X100 (1%). Kinetic analysis and starch-adsorption assay indicated that the N-terminal domain of OPMA-N could increase the OPMA-N-starch binding and subsequently increase the catalytic efficiency of OPMA-N for starch. In particular, the N-terminal domain of OPMA-N did not determine its oligomerization, because both OPMA-N and ΔOPMA-N could exist in the forms of monomer, homodimer, and homooligomer at the same time.