Merck
Isolation, identification and degradation characterization of a p-chloroaniline degrading strain.
Bulletin of environmental contamination and toxicology (2011-03-02)
Cheng Ding, Zhao-xia Li, Jin-long Yan
ABSTRAKT
A bacterium (strain ycsd02) was isolated from rhizosphere soil in phragmites wetland using the enrichment culture contaminated with p-Chloroaniline (p-CA). Morphology, physio-biochemical characteristics and phylogenetic analysis based on 16S rDNA sequence indicated that the shape of the single cell is pole, which clearly belongs to Bacillus licheniformis. The optimal conditions for satisfactory degradation of p-CA were also studied. A dynamic test indicated that the biodegradation had no lag phase at concentrations from 0 to 20 mg/L in minimal medium culture fluids at vaccination of 15%, pH 7.0 and 32°C, and the growth of strain ycsd02 cells met the first-order kinetics.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
4-Chloroaniline, PESTANAL®, analytical standard
Sigma-Aldrich
4-Chloroaniline, purified by sublimation, ≥99%
Sigma-Aldrich
4-Chloroaniline, 98%
Supelco
p-Chloroaniline, Pharmaceutical Secondary Standard; Certified Reference Material