Merck
Distribution of n-octadecenoic fatty acids in triacylglycerols of ripening sea buckthorn fruit mesocarp.
Doklady. Biochemistry and biophysics (2011-03-04)
V D Tsydendambaev, E I Kuznetsova, V P Pchelkin, A G Vereshchagin
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
cis-Vaccenic acid, ≥97% (capillary GC), oil