Merck
Response to a third-party evaluation of Clinitest® hCG sensitivity to various isoforms of human chorionic gonadotropin.
Expert review of molecular diagnostics (2011-03-17)
Eric S Cowden, Gary H Krauth, David J Ledden
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Benedict′s reagent, for the qualitative determination of sugars