Merck
  • Ultrafiltration membranes composed of highly cross-linked cationic polymer gel: the network structure and superior separation performance.
Ultrafiltration membranes composed of highly cross-linked cationic polymer gel: the network structure and superior separation performance.
Advanced materials (Deerfield Beach, Fla.) (2011-03-18)
Qifeng Wang, Sadaki Samitsu, Izumi Ichinose
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Poly(4-vinylpyridine), average Mw ~160,000
Sigma-Aldrich
Poly(4-vinylpyridine), average Mw ~60,000