Merck
Frequency dependence of alternating current electrospray ionization mass spectrometry.
Analytical chemistry (2011-03-23)
Nishant Chetwani, Catherine A Cassou, David B Go, Hsueh-Chia Chang
ABSTRAKT
The novel effects resulting from the entrainment of low mobility ions during alternating current (ac) electrospray ionization are examined through mass spectrometry and voltage/current measurements. Curious phenomena such as pH modulation at high frequencies (>150 kHz) of an applied ac electric field are revealed and explained using simple mechanistic arguments. Current measurements are utilized to supplement these observations, and a simplified one-dimensional transient diffusion model for charge transport is used to arrive at a scaling law that provides better insight into the ac electrospray ionization process. Moreover, because of the different pathway for ion formation in comparison to direct current (dc) electrospray, ac electrospray (at frequencies >250 kHz) is shown to reduce the effects of ionization suppression in a mixture of two molecules with different surface activities.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Tetrapentylammonium bromide, ≥99%
Sigma-Aldrich
Tetrapentylammonium hydroxide solution, ~20% in H2O (T)
Supelco
Tetrapentylammonium bromide, suitable for ion pair chromatography, LiChropur, ≥99.0% (AT)