Merck
[Enzyme immunassay of alternariol for the assessment of risk of agricultural products contamination].
Prikladnaia biokhimiia i mikrobiologiia (2011-03-30)
A A Burkin, G P Kononenko
ABSTRAKT
A highly specific test system based on polyclonal rabbit antibodies targeting a conjugate of alternariol and bovine serum albumin prepared by formaldehyde condensation has been developed for the first time and shown to have the sensitivity of 0.4 ng/ml in the indirect competitive determination of alternariol. The possibility of applying the assay for assessment of the intensity of alternariol contamination of corn, animal feed, and raw materials used for the preparation of the latter has been demonstrated.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Alternariol, analytical standard
Sigma-Aldrich
Alternariol from Alternaria sp., ~96%