Merck
Application of sulphanilamides disazo dyes with Tropaeolin O for simple, rapid and sensitive spectrophotometric assay of medicines.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2011-04-05)
Maria Boiko, Teodoziya Vrublevska, Olha Korkuna, Grigory Teslyar
ABSTRAKT
A rapid, simple and sensitive spectrophotometric method for the determination of some sulphanilamides is described. The method is based on the formation of blue coloured disazo dyes by the diazotization of sulphonamides viz. sulphanilamide (SA), sulphamerazine (SMR), sulphamethazine (SMZ), sulphadimethoxine (SDM), sulphamethoxazole (SMX), sulphadiazine (SDA), sulfathiazole (STZ), sulphaguanidine (SGN), sulphamonomethoxine (SMM), sulphamethoxypyridazine (SMP) in 0.5M hydrochloric acid media at ice bath followed by the azocoupling reaction with acid monoazo dye Tropaeolin O (TrO) at pH=10.5. Formed products are stable for 10h at room temperature. Effective molar absorptivities at absorbance maxima 595nm for disazo dyes were ∼10(4)M(-1)cm(-1). Stoichiometric ratios of the components of disazo dyes were determined by means of mole ratio and continuous variations methods. Linear ranges for sulphanilamides determination were 0.4-14.0μgml(-1). The methods were successfully approved at suphanilamides determination in model solutions and commercial pharmaceutical preparations.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Sulfanilamide melting point standard, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sulfanilamide, puriss. p.a., ≥98% (calc. to the dried substance)
Supelco
Sulfanilamide, VETRANAL®, analytical standard
Sigma-Aldrich
Sulfanilamide, ≥98%