Merck
Use-dependent AMPA receptor block reveals segregation of spontaneous and evoked glutamatergic neurotransmission.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2011-04-08)
Yildirim Sara, Manjot Bal, Megumi Adachi, Lisa M Monteggia, Ege T Kavalali
ABSTRAKT
Earlier findings had suggested that spontaneous and evoked glutamate release activates non-overlapping populations of NMDA receptors. Here, we evaluated whether AMPA receptor populations activated by spontaneous and evoked release show a similar segregation. To track the receptors involved in spontaneous or evoked neurotransmission, we used a polyamine agent, philanthotoxin, that selectively blocks AMPA receptors lacking GluR2 subunits in a use-dependent manner. In hippocampal neurons obtained from GluR2-deficient mice, philanthotoxin application decreased AMPA-receptor-mediated spontaneous miniature EPSCs (AMPA-mEPSCs) down to 20% of their initial level within 5 min. In contrast, the same philanthotoxin application at rest decreased the subsequent AMPA-receptor-mediated evoked EPSCs (eEPSCs) only down to 80% of their initial value. A 10-min-long perfusion of philanthotoxin further decreased AMPA-eEPSC amplitudes to 60% of their initial magnitude, which remained substantially higher than the level of AMPA-mEPSC block achieved within 5 min. Finally, stimulation after removal of philanthotoxin resulted in reversal of AMPA-eEPSC block, verifying strict use dependence of philanthotoxin. These results support the notion that spontaneous and evoked neurotransmission activate distinct sets of AMPA receptors and bolster the hypothesis that synapses harbor separate microdomains of evoked and spontaneous signaling.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Lidocaine N-ethyl bromide, analytical standard, for drug analysis