Merck
Gastrointestinal dysfunction following hemolytic uremic syndrome.
Digestive diseases and sciences (2011-05-12)
Cayley Bowles, Megan Ancker, George Triadafilopoulos
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Clidinium bromide, analytical standard