Merck
Glabridin protects osteoblastic MC3T3-E1 cells against antimycin A induced cytotoxicity.
Chemico-biological interactions (2011-05-31)
Eun Mi Choi
ABSTRAKT
Mitochondrial dysfunction, particularly respiratory chain disruption, is often responsible for aging-related bone diseases. In this study, the protective effects of glabridin, an isoflavan isolated from licorice root, against pharmacological inhibition of the respiratory chain were studied using osteoblastic MC3T3-E1 cells treated with antimycin A, which inhibits complex III of the electron transport system. Glabridin restored mitochondrial membrane potential dissipation, ATP loss, inactivation of complex IV, intracellular calcium elevation, and cytochrome c release that was induced by antimycin A treatment. This compound also prevented cell death. These results imply that glabridin protects osteoblasts from antimycin A-induced cell death via improved mitochondrial function. Glabridin scavenged ROS and mitochondrial superoxide anions generated by antimycin A. In addition, glabridin prevented antimycin A-induced nitrotyrosine increase and thioredoxin reductase inactivation, suggesting that glabridin may be useful for protecting mitochondria against a burst of oxidative stress. Since phosphoinositide 3-kinase (PI3K) and cAMP-response element-binding protein (CREB) signaling is known to be pro-survival, we determined whether PI3K and CREB activation is associated with the cytoprotective effects of glabridin in the MC3T3-E1 cells. Glabridin restored antimycin A-induced inactivation of PI3K and CREB, suggesting that PI3K and CREB-dependent pathways may be involved in glabridin-induced cytoprotective responses. Our study demonstrates that glabridin reduces mitochondrial dysfunction induced during aging, and could significantly prevent osteoblast damage in osteoporotic patients.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Glabridin, ≥98% (HPLC), powder