Merck
Synthesis, characterization, tautomerism and theoretical study of some new Schiff base derivatives.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2011-06-07)
Gülşen Türkoğlu, Halil Berber, Hakan Dal, Cemil Öğretir
ABSTRAKT
New Schiff base derivatives were prepared by the condensation of 5-chloro and 5-bromo salicylaldehyde with bis(o-aminophenol)ethers. Five bis(o-nitrophenol)ether compounds were synthesized using some ditosylate, 1,3-dibromopropane and 1,4-dibromobuthane with o-nitrophenol. These compounds were reduced to bis(o-aminophenol)ethers. The products have been characterized by elemental analysis, FTIR, 1H, 13C NMR, HETCOR and HMBC spectroscopic techniques. The tautomerisms of all of the Schiff bases compounds were determined in DMSO, CHCl3, C2H5OH and C6H12 solvents and in both acidic and basic media using the UV-vis spectrophotometric method. The heat of formation (ΔHf), enthalpy (ΔH), entropy (ΔS), Gibbs free energy (ΔGf and ΔG), stable isomers, conformations and tautomers of the synthesized compounds are calculated using the MOPAC2009 (PM6) program.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
2-Aminophenol, 99%