Merck
  • The next chapter in MOF pillaring strategies: trigonal heterofunctional ligands to access targeted high-connected three dimensional nets, isoreticular platforms.
The next chapter in MOF pillaring strategies: trigonal heterofunctional ligands to access targeted high-connected three dimensional nets, isoreticular platforms.
Journal of the American Chemical Society (2011-06-17)
Jarrod F Eubank, Lukasz Wojtas, Matthew R Hight, Till Bousquet, Victor Ch Kravtsov, Mohamed Eddaoudi
ABSTRAKT
A new pillaring strategy, based on a ligand-to-axial approach that combines the two previous common techniques, axial-to-axial and ligand-to-ligand, and permits design, access, and construction of higher dimensional MOFs, is introduced and validated. Trigonal heterofunctional ligands, in this case isophthalic acid cores functionalized at the 5-position with N-donor (e.g., pyridyl- or triazolyl-type) moieties, are designed and utilized to pillar pretargeted two-dimensional layers (supermolecular building layers, SBLs). These SBLs, based on edge transitive Kagomé and square lattices, are cross-linked into predicted three-dimensional MOFs with tunable large cavities, resulting in isoreticular platforms.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Isophthalic acid, 99%