Merck
Genotoxic effects of chlorophenoxy herbicide diclofop-methyl in mice in vivo and in human lymphocytes in vitro.
Drug and chemical toxicology (2011-07-01)
Fatma Ünal, Deniz Yüzbaşıoğlu, Serkan Yilmaz, Nihan Akinci, Hüseyin Aksoy
ABSTRAKT
Diclofop-methyl (DM) is a chlorophenoxy derivative used in large quantities for the control of annual grasses in grain and vegetable crops. In this study, the genotoxic effects of DM were investigated by measuring chromosomal aberrations (CAs) in mouse bone-marrow cells and CA and the comet assay in human peripheral lymphocytes. Mice were treated with 15.63, 31.25, 62.5, and 125 mg/kg body weight of DM intraperitoneally for 24 hours, and 15.63-, 31.25-, 62.5-, 125-, and 250-µg/mL concentrations were applied to human lymphocytes for both 24 and 48 hours. In in vivo treatments, DM significantly, but not dose dependently, increased the total chromosome aberrations, compared to both negative and solvent controls. Cell proliferation was significantly, but not dose dependently, affected by all doses. In in vitro treatments, DM (except 15.63 µg/mL) significantly and dose dependently increased the frequency of chromosome aberrations. Also, 250 µg/mL of 48-hour treatment was found to be toxic. Cell proliferation was significantly and dose dependently affected by DM applications, when compared to negative control. In in vitro treatments, DM significantly decreased the mitotic index only at the highest concentration for 24 hours, and 62.5- and 125-µg/mL concentrations for 48 hours. In the comet assay, a significant and dose-dependent increase in comet-tail intensity was observed at 62.5-, 125-, and 250-µg/mL concentrations. The mean comet-tail length was significantly increased in all concentrations. Our results demonstrate that DM is genotoxic in mammalian cells in vivo and in vitro.