Merck
  • HPLC determination of fumonisin mycotoxins in maize: a comparative study of naphthalene-2,3-dicarboxaldehyde and o-phthaldialdehyde derivatization reagents for fluorescence and diode array detection.
HPLC determination of fumonisin mycotoxins in maize: a comparative study of naphthalene-2,3-dicarboxaldehyde and o-phthaldialdehyde derivatization reagents for fluorescence and diode array detection.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2011-07-06)
Ncediwe Ndube, Liana van der Westhuizen, Ivan R Green, Gordon S Shephard
ABSTRAKT
Fumonisins are mycotoxins produced by various species of Fusarium and occur naturally in contaminated maize and maize-based foods. Ingestion of fumonisins has considerable health implications for humans and animals. Since fumonisins lack a useful chromophore or fluorophore, their determination in maize is routinely achieved via HPLC with fluorescence detection (FLD) after precolumn derivatization. This study optimized naphthalene-2,3-dicarboxaldehyde (NDA) derivatization of fumonisins in naturally contaminated maize following strong anion exchange (SAX) solid phase extraction (SPE) clean-up and utilizing diode array detection (DAD) as a practical alternative simultaneously to FLD. The limit of detection (LOD) for fumonisin B(1) (FB(1)), fumonisin B(2) (FB(2)) and fumonisin B(3) (FB(3)) with FLD was 0.11 ng, 0.50 ng and 0.27 ng, respectively, and with DAD it was 13.8 ng, 12.5 ng and 6.6 ng, respectively injected on column. The coefficient of variation (CV, n = 6) for FB(1), FB(2) and FB(3) in a naturally contaminated samples obtained with FLD was 2.6%, 1.8% and 5.3%, respectively, compared to 6.0%, 3.4% and 9.5%, respectively, obtained with DAD. Subsequently the optimized NDA derivatization was compared to the widely used o-phthaldialdehyde (OPA) derivatization agent as well as alternative sample clean-up with immunoaffinity column (IAC) by analyzing naturally contaminated maize samples (n = 15) ranging in total fumonisin (TFB = FB(1)+FB(2)+FB(3)) levels from 106 to 6000 μg/kg. After immunoaffinity column clean-up of extracted samples, the recoveries of spiked maize samples for NDA-FLD of FB(1), FB(2) and FB(3) were 62%, 94% and 64%, respectively. NDA proved to be an effective derivatization reagent of fumonisin in naturally contaminated maize samples following IAC clean-up, except for DAD at TFB levels below 1000 μg/kg. In contrast NDA derivatization following SAX clean-up produced results comparable to OPA only for levels below 1000 μg/kg. Aside from the difference in detection limits, FLD and DAD produced comparable results irrespective of the clean-up method or the derivatization agent.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
2,3-Naphthalenedicarboxaldehyde, suitable for fluorescence