Merck
  • Functionalizing metal nanostructured film with graphene oxide for ultrasensitive detection of aromatic molecules by surface-enhanced Raman spectroscopy.
Functionalizing metal nanostructured film with graphene oxide for ultrasensitive detection of aromatic molecules by surface-enhanced Raman spectroscopy.
ACS applied materials & interfaces (2011-07-07)
Xiaojuan Liu, Linyuan Cao, Wei Song, Kelong Ai, Lehui Lu
ABSTRAKT
Surface-enhanced Raman spectroscopy (SERS) as a powerful analytical tool has gained extensive attention. Despite of many efforts in the design of SERS substrates, it remains a grand challenge for creating a general substrate that can detect diverse target analytes. Herein, we report our attempt to address this issue by constructing a novel metal-graphene oxide nanostructured film as SERS substrate. Taking advantages of the high affinity of graphene oxide (GO) toward aromatic molecules and the SERS property of nanostructured metal, this structure exhibits great potential for diverse aromatic molecules sensing, which is demonstrated by using crystal violet (CV) with positive charge, amaranth with negative charge, and neutral phosphorus triphenyl (PPh(3)) as model molecules.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Amaranth, analytical standard
Sigma-Aldrich
Amaranth, Dye content 85-95 %