Merck
  • Determination of aliphatic amines by gas chromatography-mass spectrometry after in-syringe derivatization with pentafluorobenzoyl chloride.
Determination of aliphatic amines by gas chromatography-mass spectrometry after in-syringe derivatization with pentafluorobenzoyl chloride.
Journal of chromatography. A (2011-07-19)
Deepesh Kumar Singh, Sunil Kumar Sanghi, Sorna Gowri, Navin Chandra, Shail Bala Sanghi
ABSTRAKT
A simple and highly sensitive gas chromatographic method has been developed for the determination of low molecular weight short-chain aliphatic amines (SCAAs) after their simultaneous extraction and in-syringe derivatization with pentafluorobenzoyl chloride (PFBOC). Derivatization of the low molecular weight aliphatic amines in bicarbonate buffer of pH 10.5 with PFBOC was followed by immersed solvent microextraction. Derivatization conditions, including reagent concentration, reaction pH, ionic concentration of buffer, reaction time, stirring rate, reaction temperature and extraction solvent, have been investigated for method optimization. Linearity was studied within range of 0.15 pg ml⁻¹-50 ng ml⁻¹. The correlation coefficients were between 0.9934 and 0.9999. Detection limit of derivatized amines proved to be in the range of 0.117-1.527 pg ml⁻¹, and the intraday and interday relative standard deviation (RSD) values were less than 8% with respect to peak area. The method was applied for analysis of lake, river and industrial waste water. The recoveries of extraction from lake, river and industrial waste water samples, which have been spiked with different levels of aliphatic amines, were in the range of 68-99%, 63-102% and 62-105%, respectively.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
2,3,4,5,6-Pentafluorobenzoyl chloride, 99%