Merck
Micelle to solvent stacking of two alkaloids in nonaqueous capillary electrophoresis.
Journal of chromatography. A (2011-07-26)
Hua-dong Zhu, Wen-juan Lü, Hong-hong Li, Yan-hua Ma, Shao-qiang Hu, Hong-li Chen, Xing-guo Chen
ABSTRAKT
This paper for the first time describes the development of micelle to solvent stacking (MSS) to nonaqueous capillary electrophoresis (NACE). In this proposed MSS-NACE, sodium dodecyl sulfate (SDS) micelles transport, release, and focus analytes from the sample solution to the running buffer using methanol as their solvent. After the focusing step, the focused analytes were separated via NACE. The focusing mechanism and influencing factors were discussed using berberine (BBR) and jatrorrhizine (JTZ) as model compounds. And the optimum condition was obtained as following: 50 mM ammonium acetate, 6% (v/v) acetic acid and 10 mM SDS in redistilled water as sample matrix, 50 mM ammonium acetate and 6% (v/v) acetic acid in pure methanol as the running buffer, -20 kV focusing voltage with 30 min focusing time. Under these conditions, this method afforded limits of detection (S/N=3) of 0.002 μg/mL and 0.003 μg/mL for BBR and JTZ, respectively. In contrast to conventional NACE, the concentration sensitivity was improved 128-153-fold.