Merck
Access to optically active 3-azido- and 3-aminopiperidine derivatives by enantioselective ring expansion of prolinols.
Organic letters (2011-07-29)
Anne Cochi, Domingo Gomez Pardo, Janine Cossy
ABSTRAKT
The activation of N-alkyl prolinols by XtalFluor E allowed the formation of an aziridinium intermediate that can react with tetrabutylammonium azide (nBu(4)NN(3)) to produce 3-azidopiperidines and/or 2-(azidomethyl)pyrrolidines, in a ratio up to 100/0. These 3-azidopiperidines can be reduced to the corresponding 3-aminopiperidines.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
(S)-(+)-2-Pyrrolidinemethanol, 97%