Merck
  • Sequential injection analysis implementing multiple standard additions for As speciation by liquid chromatography and atomic fluorescence spectrometry (SIA-HPLC-AFS).
Sequential injection analysis implementing multiple standard additions for As speciation by liquid chromatography and atomic fluorescence spectrometry (SIA-HPLC-AFS).
Talanta (2011-08-03)
José P Jesus, Carlos A Suárez, José R Ferreira, Maria F Giné
ABSTRAKT
An analytical procedure for multiple standard additions of arsenic species using sequential injection analysis (SIA) is proposed for their quantification in seafood extracts. SIA presented flexibility for generating multiple specie standards at the ng mL(-1) concentration level by adding different volumes of As(III), As(V), monomethylarsonic (MMA) and dimethylarsinic (DMA) to the sample. The mixed sample plus standard solutions were delivered from SIA to fill the HPLC injection loop. Subsequently, As species were separated by HPLC and analyzed by atomic fluorescence spectrometry (AFS). The proposed system comprised two independently controlled modules, with the HPLC loop acting as the intermediary device. The analytical frequency was enhanced by combining the actions of both modules. While the added sample was flowing through the chromatographic column towards the detection system, the SIA program started performing the standard additions to another sample. The proposed method was applied to spoiled seafood extracts. Detection limits based on 3σ for As(III), As(V), MMA and DMA were 0.023, 0.39, 0.45 and 1.0 ng mL(-1), respectively.