Merck
  • Palladium-catalyzed stereoconvergent formylation of (E/Z)-β-bromo-β-fluorostyrenes: straightforward access to (Z)-α-fluorocinnamic aldehydes and (Z)-β-fluorocinnamic alcohols.
Palladium-catalyzed stereoconvergent formylation of (E/Z)-β-bromo-β-fluorostyrenes: straightforward access to (Z)-α-fluorocinnamic aldehydes and (Z)-β-fluorocinnamic alcohols.
The Journal of organic chemistry (2011-08-05)
Rajae Zemmouri, Mohammed Kajjout, Yves Castanet, Said Eddarir, Christian Rolando
ABSTRAKT
We report here the stereoconvergent formylation of (E/Z)-β-bromo-β-fluorostyrene mixtures with carbon monoxide and sodium formate catalyzed by palladium. Optimization of reaction conditions leads to the corresponding pure (Z)-α-fluorocinnamaldehydes in good yields. The reaction was extended to styrenes bearing electro-attracting or electro-donating groups. The obtained α-fluoroaldehydes were smoothly reduced to the corresponding (Z)-β-fluorocinnamic alcohol by NaBH(4). The reaction could be performed on functionalized substrates as demonstrated by the access to the glucoside of β-fluoroconiferyl alcohol, (Z)-β-fluoroconiferin, a strong inhibitor of lignin polymerization.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Cinnamyl alcohol, 98%
Sigma-Aldrich
Cinnamyl alcohol, ≥98%, FG