Merck
  • The feasibility of growing cells of Saccharomyces cerevisiae for citronellol production in a continuous-closed-gas-loop bioreactor (CCGLB).
The feasibility of growing cells of Saccharomyces cerevisiae for citronellol production in a continuous-closed-gas-loop bioreactor (CCGLB).
Bioresource technology (2011-08-13)
Aimi Aishah Arifin, Mashitah Mat Don, Mohamad Hekarl Uzir
ABSTRAKT
The present work aims to address the gas-phase biotransformation of geraniol into citronellol using growing cells of Saccharomyces cerevisiae (baker's yeast) in a continuous-closed-gas-loop bioreactor (CCGLB). This study revealed that the gaseous geraniol had a severe effect on the production of biomass during the growing cell biotransformation resulting in the decrease in the specific growth rate from 0.07 to 0.05 h⁻¹. The rate of reaction of the growing cell biotransformation was strongly affected by agitation and substrate flow rates. The highest citronellol concentration of 1.18 g/L and initial rate of reaction of 7.06 × 10⁻⁴ g/min g(cell) were obtained at 500 rpm and 8 L/min, respectively.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
(−)-β-Citronellol, analytical standard
Sigma-Aldrich
(R)-(+)-β-Citronellol, 95%
Sigma-Aldrich
(S)-(−)-β-Citronellol, ≥97%
Sigma-Aldrich
Citronellol, ≥95%, FCC, FG
Sigma-Aldrich
β-Citronellol, 95%
Sigma-Aldrich
Citronellol, mixture of isomers, natural, ≥95%, FG
Supelco
(±)-β-Citronellol, analytical standard