Merck
Methods for determination of all binding parameters in systems with simultaneous borate and cyclodextrin complexation.
Journal of chromatography. A (2011-08-30)
Jana Svobodová, Pavel Dubský, Eva Tesařová, Martin Beneš, Bohuslav Gaš
ABSTRAKT
Two novel methods for determination of binding constants in the systems with borate and cyclodextrin complexation were developed. The methods enable to determine all binding parameters in these systems and even the binding constants of interaction of a neutral analyte with a neutral cyclodextrin. The first method is based on nonlinear fitting of experimental data and further evaluation of fitting parameters. The second method requires a multiple regression. The methods provide identical results with low experimental error. Only one set of measurements is required for both methods. Thus the binding parameters can be mutually compared. The binding parameters for neutral analytes ((R,R)-(+)-hydrobenzoin and (S,S)-(-)-hydrobenzoin) and neutral cyclodextrin (heptakis(2,6-di-O-methyl)-β-cyclodextrin) were evaluated and the effect of individual types of interaction was revealed. The interaction of the analytes with cyclodextrin governs the chiral recognition, while the complexation of analyte with borate is responsible for electromigration. Very low values of the binding constants of mixed analyte-cyclodextrin-borate complexes indicate that this type of complexation has negligible effect on enantioseparation.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Benzoin, purified by sublimation, ≥99.5%
Sigma-Aldrich
Benzoin, 98%
Sigma-Aldrich
Heptakis(2,6-di-O-methyl)-β-cyclodextrin