Merck
Synthesis of substituted [3]dendralenes and their unique cycloaddition reactions.
Chemical communications (Cambridge, England) (2011-08-30)
Rekha Singh, Sunil K Ghosh
ABSTRAKT
Dimethylsulfonium methylide mediated olefination of 2-phenylethenylidene phosphonoacetate followed by the Horner-Wadsworth-Emmons reaction with aromatic aldehydes provided access to reactive 1,5-diaryl-2-ethoxycarbonyl [3]dendralenes which in situ underwent Diels-Alder cyclodimerisation leading to highly functionalised cyclohexenes with very high regio- and stereoselectivity.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Cyclohexene, inhibitor-free, ReagentPlus®, 99%
Sigma-Aldrich
Cyclohexene, contains 100 ppm BHT as inhibitor, ≥99.0%
Supelco
Cyclohexene, analytical standard, ≥99.5% (GC)