Merck
Enzyme-assisted synthesis of the glucuronide conjugate of psilocin, an hallucinogenic component of magic mushrooms.
Drug testing and analysis (2011-10-01)
Takuji Shoda, Kiyoshi Fukuhara, Yukihiro Goda, Haruhiro Okuda
ABSTRAKT
An enzyme-assisted synthesis of psilocin glucuronide (PCG), a metabolite excreted in the urine of magic mushroom (MM) users, is described. In the presence of Aroclor 1254 pretreated rat liver microsomes, psilocin and the cofactor UDPGA were incubated for 20 h. Purification by HPLC gave PCG in 19% yield (3.6 mg). The compound structure was characterized by MS and NMR. The milligram amounts of PCG produced by this method will allow the direct identification and quantification of PCG in the urine of MM users.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Aroclor 1254, ampule of 50 mg, analytical standard
Supelco
Aroclor 1254 solution, certified reference material, 500 mg/kg in transformer oil
Supelco
Aroclor 1254 solution, certified reference material, 50 mg/kg in transformer oil